Would you like to log in again?
NAD Nucleotidase (from E. coli)
Supplier: AdipoGen Life Sciences
AG-40T-0110-C050
102980-456EA 540 USD InStock
102980-456
NAD Nucleotidase (from E. coli)
Enzymes
NadN (NAD nucleotidase) is a periplasmic enzyme from Haemophilus influenzae, a major pathogen of the respiratory tract in humans that has developed the capability to exploit host NAD(P) for its nicotinamide dinucleotide requirement
  • UniProt link Q4QNY4: NAD Nucleotidase (H. influenza)
NadN plays a central role by degrading NAD into adenosine and NR (nicotinamide riboside), NMN to nicotinamide riboside and AMP to adenosine. It shows a broad substrate specificity, recognizing either mono- or di-nucleotide nicotinamides and different adenosine phosphates with a maximal activity on 5'-adenosine monophosphate. It can be used as an economically alternative to produce NR (nicotinamide riboside). Catalytic activity: NAD=AMP + NMN=Ade+NR+PPi | AMP=Ade+Pi | NMN=NR+Pi
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty