Would you like to log in again?
Human Recombinant EPO (from CHO cells)
Supplier: AdipoGen Life Sciences
CHI-HF-220EPO-C050
102981-552EA 498.46 USD InStock
102981-552
Human Recombinant EPO (from CHO cells)
Proteins and Peptides
Erythropoietin is the principal hormone involved in the regulation of erythropoiesis by stimulating the proliferation and differentiation of erythroid progenitor cells and the maintenance of a physiological level of circulating erythrocyte mass. It is a glycoprotein produced primarily by the kidney. The biological effects of EPO are mediated by the erythropoietin receptor (Epo R). Genetic variations in EPO is associated with susceptibility to microvascular complications of diabetes type 2 (MVCD2) (including diabetic retinopathy, diabetic nephropathy leading to end-stage renal disease and diabetic neuropathy). It is used for the treatment of anemia and misused as a performance-enhancing drug in endurance athletes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty