Would you like to log in again?
Penicillium Citrinum Nuclease P1
Supplier: AdipoGen Life Sciences
JAI-OPNUCP1
102981-726EA 2690.31 USD InStock
102981-726
Penicillium Citrinum Nuclease P1
Enzymes
Nuclease P1 is a zinc dependent glycoprotein that catalyzes the nonspecific endonucleolytic cleavage of single stranded DNA and RNA to yield nucleoside 5'-phosphates and 5'-phospho-oligonucleotides.
  • Consists of 270 amino acid residues
  • Optimal pH: 5.3 (RNA and heat-denatured DNA); 7.2 (3'-AMP)
  • Stable below 60°C at pH 5.3 for 30 minutes
  • Used for DNA samples pretreatment to detect 8-OHdG
Nuclease P1 hydrolyzes both 3'-5'-phosphodiester bonds in RNA and heat-denatured DNA and 3'-phosphomonoester bonds in mono- and oligonucleotides terminated by 3'-phosphate without base specificity
One unit of this enzyme will liberate
1.0 μmol of acid soluble nucleotides from RNA per min at pH 5.3 at 37°C. 3'-Nucleotidase and will hydrolyze
4.0 μmol of orthophosphate from 3'-AMP per min at pH 7.2 at 37°C. This enzyme has an optimal temperature of approximately 70°C, but for a long incubation, a temperature below 60°C is more suitable. It is stable in the pH range of 5 to 8.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty