Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2,4-Diaminobutanoic acid dihydrochloride

Supplier: ADIPOGEN CORP MS
Synonyms: H-Dab-OH

CDX-D0154-G005 CDX-D0154-G001
102987-908EA 279.69 USD InStock
102987-908 102987-906
(S)-2,4-Diaminobutanoic acid dihydrochloride
(S)-2,4-Diaminobutanoic acid dihydrochloride
Unnatural amino acid derivative. Pharmacological tool and potential chiral building block used as an internal standard for amino acid analysis. Suitable reagent used for the differentiation of beta-N-methylamino-L-alanine from the diamino acids by using HPLC-FD, UHPLC-UV, UHPLC-MS, and triple quadrupole tandem mass spectrometry (UHPLC-MS/MS). Used in the quantification of neurotoxin beta-N-methylamino-L-alanine (BMAA) in seafood. It has been found to inhibit GABA transaminase (ABAT), producing elevated levels of GABA and to have an antitumor activity.


Formula: C4H12Cl2N2O2
MW: 191.06 g/mol
MDL Number: MFCD01632031
CAS Number: 1883-09-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty