Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-Platin

Supplier: ADIPOGEN CORP MS
Synonyms: cis-Diamminedichloroplatinum (II) cis-Diammineplatinum (II) dichloride Platinol

AG-CR1-3590-M250 AG-CR1-3590-M050
102988-682EA 96.92 USD InStock
102988-682 102988-680
cis-Platin
cis-Platin
Potent platinum-based antineoplastic agent. Anticancer compound. Forms inter- and intrastrand DNA adducts/crosslinks, consequently blocking DNA replication and transcription and inducing cell death. Nephrotoxic when used in vivo. Potent chemoimmunotherapeutic. Stimulates the immune responses by activating macrophages and other cells of the immune system. Apoptosis inducer via p53-dependent and -independent mechanisms. Induces apoptosis through caspase-3 activation and XIAP expression inhibition. Targets other proteins and enzymes. Non-competitive NHE-1 inhibitor, an archetypal, membrane-bound mechanosensitive sodium-hydrogen ion transporter.


Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
MDL Number: MFCD00011623
CAS Number: 15663-27-1
UN: 3288
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty