Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Melanin Production Inhibitor A3B5 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning
Synonyms: N-(3,5-Dimethylphenyl)-3-methoxybenzamide

AG-CR1-3605-M001
102988-740EA 166.15 USD InStock
102988-740
Melanin Production Inhibitor A3B5 ≥98% (by NMR)
Melanin production inhibitor A3B5
Non-cytotoxic melanogenesis inhibitor. Potential depigmentation agent. Tyrosinase-related protein 2 (TRP-2) expression inhibitor, via proteasomal degradation. Inhibits the proliferation of melanocytes and melanoma cells through cell cycle arrest in G1. Moderate skin tumor growth suppressor in vivo.


Formula: C₁₆H₁₇NO₂
MW: 255.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01152934
CAS Number: 346721-60-6
Order Now
Specification Test Results
Purity 98% (NMR)
Appearance/Color White solid.
Solubility Soluble in DMSO or methanol. Insoluble in water.

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty