Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Empagliflozin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning
Synonyms: Jardiance(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

AG-CR1-3619-M010 AG-CR1-3619-M050
102988-794EA 249.23 USD InStock
102988-794 102988-792
Empagliflozin ≥98% (by HPLC)
Empagliflozin
Antidiabetic agent. Potent and selective sodium glucose co-transporter 2 (SGLT-2) inhibitor. 300-fold selectivity over SGLT-1, 4, 5 and 6. SGLT-2 is found almost exclusively in the proximal tubules of nephronic components in kidneys. Inhibition of SGLT-2 reduces blood glucose by blocking renal glucose reabsorption and thereby increasing urinary glucose excretion (UGE). Shown to preserves beta cell mass and restore glucose homeostasis.


Formula: C₂₃H₂₇ClO₇
MW: 450.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22566222
CAS Number: 864070-44-0
Order Now
Specification Test Results
Purity 98% (HPLC)
Appearance/Color White solid.
Solubility Soluble in DMSO. Slightly soluble in water or ethanol.

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty