Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',7'-Dichlorofluorescein

Supplier: ADIPOGEN CORP MS
Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

CDX-D0004-G010 CDX-D0004-G025 CDX-D0004-G100
102989-600EA 139.85 USD InStock
102989-600 102989-602 102989-604
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein
Fluorescent dye. Spectral data: Extinction 504nm/Emmission 529nm, in 0.1 M Tris pH 8.0. Used for the determination of carbohydrates by fluorescence densitometry after TLC, in the determination of H2O2 by chemiluminescence, and as a fluorescent additive used in indirect fluorimetric detection in OTC-HPLC. Used as a component/substrate of the 2′,7′-dichlorofluorescein diacetate (DCFH-DA) assay to quantitate reactive oxygen species (ROS).


Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
MDL Number: MFCD00005047
CAS Number: 76-54-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty