Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

Supplier: ADIPOGEN CORP MS
Synonyms: 3,3'-Dithiobis(6-nitrobenzoic acid) Ellman's reagent 2,2'-Dinitro-5,5'-dithiodibenzoic acid DTNB

CDX-D0156-G001 CDX-D0156-G005 CDX-D0156-G025
102989-640EA 96.92 USD InStock
102989-640 102989-642 102989-638
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
A sensitive reagent for measuring the free sulfhydryl content in proteins, peptides, and tissues. Used to characterize reactive thiol groups and photometric determination of thiols and for measuring low-molecular mass thiols such as glutathione in both pure solutions and biological samples, such as blood. It can also measure the number of thiol groups on proteins. Through reaction with aliphatic thiol groups a mixed disulfide of protein thiol and one mole of 2-nitro-5-thiobenzoate per mole of protein sulfhydryl group is being formed. DTNB has little absorbance. Reaction with -SH groups on proteins (from any solvent accessible Cys) under mild alkaline conditions (pH 7-8) produces the 2-nitro-5-thiobenzoate anion, which gives an intense yellow color with an absorption maximum at 409.5nm (Extinction coefficient: 14150 M-1*cm-1). Sensitive to various buffer ions, therefore, the extinction coefficient used to calculate the number of sulfhydryl groups must be matched to the reaction conditions. In case the thiol groups are in disulfide bonds, they must be reduced under anaerobic conditions prior to reaction with DTNB.


Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
MDL Number: MFCD00007140
CAS Number: 69-78-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty