Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DAABD-AE ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Synonyms: 4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole

CDX-D0204-M050 CDX-D0204-M250
102989-660EA 333.69 USD InStock
102989-660 102989-662
DAABD-AE ≥95% (by HPLC)
DAABD-AE
4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole (DAABD-AE) is new derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids (i.e. hexacosanoic, tetracosanoic, and docosanoic acids) in plasma.


Formula: C₁₂H₂₀N₆O₃S
MW: 328.39 g/mol
Storage Temperature: Freezer
CAS Number: 913253-56-2
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Orange powder.
Solubility Soluble in chloroform.

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty