Would you like to log in again?
Isoprostane EIA Kit, Eagle Biosciences
Supplier: Eagle Biosciences
15I39-K01
102993-336EA 712.69 USD InStock
102993-336
Isoprostane EIA Kit, Eagle Biosciences
Assays ELISAs
Isoprostane ELISA is to detect 15-Isoprostane F2t in biological fluids by a microplate ELISA.
  • Dynamic Range: 0.05 - 100 ng/mL
  • Sensitivity: 0.05 ng/mL
  • Incubation: 2.5 hours
Isoprostanes are prostaglandin-like compounds that are produced by free radical mediated peroxidation of lipoproteins. This kit is for the quantification of a representative isoprostane, 15-isoprostane F2t (also known as 8-epi-PGF2a, 8-iso-PGF2a or 8-isoprostane). Levels of 15-isoprostane F2t (15-IsoP F2t) in biological fluids have been shown to be useful for assessment of oxidant stress in vivo. 15-IsoP F2t has also been shown to be a potent vasoconstrictor in rat kidneys and rabbit lungs, and plays a causative role in atherogenesis. Elevated isoprostane levels are associated with hepatorenal syndrome, rheumatoid arthritis, atherosclerosis, and carcinogenesis. The 15-Isoprostane F2t ELISA kit can be used for the quantification of free 15-IsoP F2t in urine, plasma, serum, and tissue samples following Solid Phase Extraction (SPE) of the isoprostane-containing fraction. Instructions are also provided for the quantification of total 15-IsoP F2t following hydrolysis of phospholipids. NOTE: Alternatively, urine samples may be analyzed without SPE using a separate kit, EA85 – Urinary Isoprostane ELISA, also available from Oxford Biomedical Research.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty