Would you like to log in again?
11 12-EET/DHET ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
11E39-K05 11E39-K02 11E39-K01 11E39-K10
102993-368EA 2875 USD InStock
102993-368 102993-362 102993-364 102993-366
11 12-EET/DHET ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The Eagle Biosciences 11,12-EET/DHET ELISA Assay kit isis intended for the quantitative determination of 11,12-EET/DHET in biological samples by ELISA and is For Research Use Only.
  • Measure 11,12-EET/DHET in biological samples
  • Can be used to measure EET levels in cultured cells which express sEH
  • Does not cross-react with 5(S)15(S)DiHETE and arachidonic acid (AA)
The Eagle Biosciences 11,12-EET/DHET ELISA Assay kit is intended for the quantitative determination of 11,12-EET/DHET in biological samples by enzyme linked immunoassay (ELISA). The Eagle Biosciences 11,12-EET/DHET ELISA Assay kit is for research use only and not to be used in diagnostic procedures
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty