Would you like to log in again?
14 15-EET/DHET ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
14E39-K10 14E39-K01 14E39-K02 14E39-K05
102993-380EA 555.77 USD InStock
102993-380 102993-382 102993-076 102993-078
14 15-EET/DHET ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The Eagle Biosciences 14,15-EET/DHET ELISA Assay kit is intended for the quantitative determination of 14,15-EET/DHET in biological samples by enzyme linked immunoassay (ELISA)
  • Measure 14,15-EET/DHET in biological samples
  • Intended for the quantitative determination of 14,15-EET/DHET
  • Competitive ELISA
The Eagle Biosciences 14,15-EET/DHET ELISA Assay kit is intended for the quantitative determination of 14,15-EET/DHET in biological samples by enzyme linked immunoassay (ELISA). The Eagle Biosciences 14,15-EET/DHET ELISA Assay kit is for research use only and not to be used in diagnostic procedures.
The Eagle Biosciences 14,15-EET/DHET ELISA Assay kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty