Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GP 2 (Glycoprotein 2)IgG ELISA, Eagle Biosciences

Supplier: Eagle Biosciences
3850
102993-394EA 568.85 USD InStock
102993-394
Anti-GP 2 (Glycoprotein 2)IgG ELISA, Eagle Biosciences
Assays ELISAs
Anti-GP2 IgG determines IgG antibodies against glycoprotein 2 in human serum.

  • Dynamic Range: 1 - 300 U/ml
  • Sensitivity: 1 U/mL
  • Incubation: 1.5 hours

Non-specific inflammatory bowel disease including Crohn’s disease (Enteritis regionalis) and ulcerative colitis (UC) are characterised by unknown etiology as well as chronic-remitting inflammatory processes of the intestine. Whereas the inflammation of ulcerative colitis is restricted to the mucosa and submucosa of colon and rectum, Crohn’s disease (CD) shows a wide spread inflammation of gastro-intestinal tract with granuloma formation. The risk developing one of these diseases is strongly correlated to immunologic, genetic, infectious and environmental factors. The differential diagnosis of inflammatory bowel diseases to chronic diarrhea, recurrent abdominal dolor, infectious colitis, anorexia as well as the differentiation of CD to UC is still a challenge. Autoantibodies of exocrine pancreas (PAB) were identified as specific serological marker for CD. A prevalence of 39 % of these autoantibodies in patients with CD could be demonstrated by indirect immune fluorescence (Stöcker et al. 1987). Glycoprotein 2 (GP2), a membrane-bound pancreatic protein, could be identified/verified as the major target of PAB’s (Roggenbuck et al., 2009). In combination with the detection of autoantibodies to Saccharomyces cerevisiae (ASCA) with a prevalence of 70 % in patients with CD and atypical antineutrophile cytoplasmatic Antigenss (aANCA) which are mainly found in patients with UC, PAB’s against GP2 could be used as a highly specific serological marker for differential diagnosis of CD to UC.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty