Would you like to log in again?
Anti-GAD (Glutamic Acid Decarboxylase) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
3802
102993-396EA 1203.08 USD InStock
102993-396
Anti-GAD (Glutamic Acid Decarboxylase) ELISA, Eagle Biosciences
Assays ELISAs
Anti-GAD (Glutamic Acid Decarboxylase) ELISA detects autoantibodies to GAD65 Abs in human serum.
  • Dynamic Range: 5 - 250 IU/ml
  • Sensitivity: 0.8 IU/ml
  • Incubation: 3 hours
The Eagle Biosciences Anti GAD ELISA Assay Kit (Enzyme immunoassay)determines autoantibodies to Glutamic Acid Decarboxylase (GAD65 Abs) in human serum. The Eagle Biosciences Anti GAD ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty