Would you like to log in again?
Gliadin IgA ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
3710
102993-402EA 568.85 USD InStock
102993-402
Gliadin IgA ELISA, Eagle Biosciences, Inc.
Assays ELISAs
GliaDea IgA ELISA determines IgA Ab against deamidated gliadin in human serum or plasma.
  • Dynamic Range: 10-300 U/mL
  • Sensitivity: 10 U/mL
  • Incubation: 2 hours
Celiac disease (CD, sprue, gluten enteropathy) is an intolerance to gluten, a cereal protein present primarily in wheat. This gluten-induced enteropathy leads to a severe damage of the small intestine and a so-called “flat” mucosa. Due to this extensive lesions mal-absorption occurs accompanied with a depletion of key nutrients.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty