Would you like to log in again?
Gliadin IgG ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
3810
102993-404EA 568.85 USD InStock
102993-404
Gliadin IgG ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Gliadin IgG ELISA is used to detect IgG antibodies against deamidated gliadin in human serum.
  • Dynamic Range: 1 - 300 U/ml
  • Sensitivity: 1 U/ml
  • Incubation: 2 hours
The Eagle Biosciences Gliadin IgG ELISA Assay Kit is used for the quantitative determination of IgG antibodies against deamidated gliadin in human serum. The Gliadin IgG ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty