Would you like to log in again?
Human IFN-g HS ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IFG39-K01
102993-428EA 1000.38 USD InStock
102993-428
Human IFN-g HS ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
The Human high sensitivity Interferon Gamma ELISA detects interferon gamma in biological samples.
  • Dynamic Range: 0.78-25 pg/ml
  • Sensitivity: 0.69 pg/ml
  • Incubation: 3h00
The Human High Sensitivity IFNγ (Interferon Gamma) ELISA Assay Kit is to be used for the in-vitro quantitative determination of interferon gamma (IFNγ) in human sera, plasmas, buffered solutions or cell culture media. The assay will recognize both natural and recombinant human IFNγ. The Eagle Biosciences Human High Sensitivity IFNγ (Interferon Gamma) ELISA Assay Kit is for Research Use Only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty