Would you like to log in again?
IgA (Canine) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
IGA49-K01
102993-438EA 1340.38 USD InStock
102993-438
IgA (Canine) ELISA, Eagle Biosciences
Assays ELISAs
Canine IgA ELISA is designed to detect canine IgA in complex samples.
  • Dynamic Range: 3.125 - 200 ng/ml
  • Sensitivity: 156 ng/ml (sample dilution 1:50)
  • Incubation: 2.5 hours
The Eagle Biosciences Canine (Dog) IgA ELISA Assay Kit is designed for the quantitative determination of canine (dog) IgA in complex samples (serum or other biological samples). The Dog IgA ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty