Would you like to log in again?
IgA (rabbit) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
IGA79-K01
102993-442EA 1340.38 USD InStock
102993-442
IgA (rabbit) ELISA, Eagle Biosciences
Assays ELISAs
Rabbit IgA ELISA is designed to detect IgA antibodies in rabbit serum and biological samples.
  • Dynamic Range: 15.625 - 1000 ng/ml
  • Sensitivity: 15.625 ng/ml
  • Incubation: 4.5 hours
The Eagle Biosciences Rabbit IgA ELISA Assay Kit is designed for the quantitative determination of IgA antibodies in rabbit serum and biological samples. The Rabbit IgA ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty