Would you like to log in again?
IgE (rat) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
IGE29-K01
102993-446EA 1399.23 USD InStock
102993-446
IgE (rat) ELISA, Eagle Biosciences
Assays ELISAs
Rat IgE ELISA is designed to detect the IgE antibodies in rat serum and biological samples.
  • Dynamic Range: 15.6 - 50 ng/ml
  • Sensitivity: 15.6 ng/ml (sample dilution 1:10)
  • Incubation: 4.5 hours
The Eagle Biosciences Rat IgE ELISA Assay Kit is designed for the quantitative determination of IgE antibodies in rat serum and biological samples. The Rat IgE ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty