Would you like to log in again?
IgG (Canine) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
IGG49-K01
102993-460EA 1340.38 USD InStock
102993-460
IgG (Canine) ELISA, Eagle Biosciences
Assays ELISAs
Canine IgG ELISA is designed to detect the IgG antibodies in canine serum and biological samples.
  • Dynamic Range: 15.625 - 1000 ng/ml
  • Sensitivity: 156 ng/ml (sample dilution 1:10)
  • Incubation: 2.5 hours
The Eagle Biosciences Canine IgG ELISA Assay Kit is designed for the quantitative determination of canine (dog) IgG in complex samples (serum or other biological samples). The Canine (dog) IgG ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty