Would you like to log in again?
IgY (chicken) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
IGG99-K01
102993-468EA 1386.15 USD InStock
102993-468
IgY (chicken) ELISA, Eagle Biosciences
Assays ELISAs
IgY (chicken) ELISA detects chicken IgY in complex samples.
  • Dynamic Range: 3.125 - 200 ng/ml
  • Sensitivity: 80 ng/ml (sample dilution 1:25)
  • Incubation: 2.5 hours
The Eagle Biosciences Chicken IgY ELISA Assay Kit is designed for the quantitative determination of chicken IgY in complex samples (serum or other biological samples). The Chicken IgY ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty