Would you like to log in again?
IgM (rat) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
IGM29-K01
102993-470EA 1340.38 USD InStock
102993-470
IgM (rat) ELISA, Eagle Biosciences
Assays ELISAs
Rat IgM ELISA is designed to detect rat IgM in complex samples (serum or other biological samples).
  • Dynamic Range: 31.25 - 2000 ng/ml
  • Sensitivity: 312.5 ng/ml (sample dilution 1:10)
  • Incubation: 2.5 hours
The Eagle Biosciences Rat IgM ELISA Assay Kit is designed for the quantitative determination of rat IgM in complex samples (serum or other biological samples). The Rat IgM ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty