Would you like to log in again?
Human IL-2 HS ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IL239-K01
102993-490EA 1000.38 USD InStock
102993-490
Human IL-2 HS ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
The Human High Sens IL-2 ELISA is for research use only and not to be used in diagnostic procedures.
  • Dynamic Range: 1.87-60 pg/ml
  • Sensitivity: 0.97 pg/ml
  • Incubation: 2h00
The Human High Sensitivity IL-2 ELISA is to be used for the in-vitro quantitative determination interleukin-2 (IL-2) in human sera, plasmas, buffered solutions or cell culture media. The assay will recognize both natural and recombinant human IL-2. The Eagle Biosciences Human High Sensitivity IL-2 ELISA Assay kit is for Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty