Would you like to log in again?
Swine IL-4 ELISA KIT, Eagle Biosciences
Supplier: Eagle Biosciences
IL451-K01
102993-502EA 660.38 USD InStock
102993-502
Swine IL-4 ELISA KIT, Eagle Biosciences
Assays ELISAs
Swine IL-4 ELISA detects Swine IL-4 concentrations in cell culture supernates, serum, and plasma.
  • Dynamic Range: 15.625 - 500 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 3.5 hours
Interleukin4 (IL-4), also known as B cell stimulatory factor1, is a monomeric, approximately 13-18 kDa Th2 cytokine that shows pleiotropic effects during immune responses (1-3).It is a glycosylated polypeptide that contains three intrachain disulfide bridges and adopts a bundled four αhelix structure (4). Porcine IL-4 is synthesized with a 24 amino acid (aa) signal sequence. Mature porcine IL-4 shares 78%, 59%, 41%, and 41% aa sequence identity with bovine, human, mouse, and rat IL-4, respectively. Human IL-4 is active on porcine vascular endothelial cells (5). IL-4 exerts its effects through two receptor complexes (6, 7). The type I receptor, which is expressed on hematopoietic cells, is a heterodimer of the ligand binding IL-4 Rα and the common γ chain (a shared subunit of the receptors for IL-2, -7, -9, -15, and -21). The type II receptor on nonhematopoietic cells consists of IL-4Rα and IL13Rα1. The type II receptor also transduces IL-13 mediated signals. IL-4 is primarily expressed by Th2-biased CD4+ T cells, mast cells, basophils, and eosinophils (1, 2). It promotes cell proliferation, survival, and immunoglobulin class switch to IgE in B cells, acquisition of the Th2 phenotype by naïve CD4+ T cells, priming and chemotaxis of mast cells, eosinophils, and basophils, and the proliferation and activation of epithelial cells (8, 11). IL-4 plays a dominant role in the development of allergic inflammation and asthma (10, 12).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty