Would you like to log in again?
Mouse IL-6 ELISA kit, Eagle Biosciences
Supplier: Eagle Biosciences
IL611-K01
102993-508EA 660.38 USD InStock
102993-508
Mouse IL-6 ELISA kit, Eagle Biosciences
Assays ELISAs
Mouse IL-6 ELISA detects mouse (IL-6) concentrations in cell culture supernates, serum, and plasma.
  • Dynamic Range: 31.25 - 1000 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 3.5 hours
The Eagle Biosciences Mouse Interleukin 6 (IL-6) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of mouse interleukin 6 (IL-6) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Mouse Interleukin 6 (IL-6) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty