Would you like to log in again?
Insulin Ultrasensitive ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
INU31-K01
102993-524EA 836.92 USD InStock
102993-524
Insulin Ultrasensitive ELISA, Eagle Biosciences, Inc.
Assays ELISAs
s Human Insulin Ultrasensitive ELISA determines human insulin in serum and/or plasma.
  • Dynamic Range: 0.145 mU/L - 7.6 mU/L
  • Sensitivity: 0.0322 mU/L
  • Incubation: 2.5 hours
Insulin is a peptide hormone, produced by beta cells of the pancreas. Enzymatic cleavage of proinsulin results in the production of equimolar amounts of insulin and C-peptide in circulation. Insulin is central to regulating carbohydrate and fat metabolism in the body.
Excessive amounts of insulin are associated with excess amounts of glucose in the system. High levels of insulin are known to cause weight gain, water bloating, high blood pressure, magnesium deficiency and an increase in the amount of inflammatory compounds in the blood, which causes blood clots and blood vessel damage.
Insulin, when insufficiently produced, results in diabetes mellitus.
In most cases, a high fasting insulin level is consistent with insulin resistance symptoms, but in some cases, it can be caused by more serious conditions such as Cushing’s syndrome, acromegaly or possibly insulinoma.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty