Would you like to log in again?
Folic Acid Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
KIFR005
102993-718EA 1870 USD InStock
102993-718
Folic Acid Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Folic acid is a microtiter plate test which measures the total folic acid content in serum.
  • Dynamic Range: 0.04 - 0.32 µg/L
  • Sensitivity: 0.04 µg/L
  • Incubation: 48 hours
The Eagle Biosciences Folic acid is a microtiter plate test kit based on a microbiological assay which measures the total folic acid content in serum.The test kit contains all required reagents,e.g. standard, medium and microtiter plate coated with a specific microorganism,sufficient for 96 determinations including standard curves.An ELISA reader is required for evaluation of the folic acid content.  The Folic Acid Assay kit is For Research Use Only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty