Would you like to log in again?
ID-Vit® Biotin Microtiter Plate Assay, Eagle Biosciences
Supplier: Eagle Biosciences
KIFR007
102993-722EA 1464.62 USD InStock
102993-722
ID-Vit® Biotin Microtiter Plate Assay, Eagle Biosciences
Assays Cellular Assays
Biotin Assay kit is a microtiter plate test kit based on a microbiological assay which measures the total free biotin content in serum.
  • Incubation Time: 48 hours
The test kit contains all required reagents, e.g. standard, medium and microtiter plate coated with a specific microorganism, sufficient for 96 determinations including standard curves. An ELISA reader is required for evaluation of the biotin con­tent. It can be used in human and veterinary medicine and in research only.
Delivery information: It includes a Lactobacillus plantarum - precoated microtiter plate (ready to use
), Water 30 mL, Biotin assay medium, Biotin standard, Cover plastic foil, Replacement holder for 96-well plates, Biotin control 1 and 2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty