Would you like to log in again?
Vitamin B12 Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
KIFR012
102993-724EA 1464.62 USD InStock
102993-724
Vitamin B12 Assay Kit, Eagle Biosciences
Assays ELISAs
Vitamin B12 test measures the total Vitamin B12 content in the serum.
  • Dynamic Range: 6 - 54 ng/L
  • Sensitivity: 6 ng/L
  • Incubation: 48 hours
The Eagle Biosciences Vitamin B12 is a microtiter plate test kit based on a microbiological assay which measures the total Vitamin B12 content in serum.The test kit contains all required reagents, e.g. standard,medium and microtiter plate coated with a specific microorganism,sufficient for 96 determinations including standard curves. An ELISA reader is required for evaluation of the Vitamin B12 content.  The Vitamin B12 Assay Kit is for research use only. 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty