Would you like to log in again?
Kallikrein-related peptidase 7 (KLK7) ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
KL731-K01
102993-726EA 1340.38 USD InStock
102993-726
Kallikrein-related peptidase 7 (KLK7) ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Kallikrein-Related Peptidase 7 (KLK7) ELISA detects the concentration of KLK7 in human serum.
  • Dynamic Range: 2 - 512 ng/ml
  • Sensitivity: 2 ng/ml
  • Incubation: 2.5 hours
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK7 is one of the fifteen kallikrein subfamily members. The KLK7 enzyme is thought to be involved in the proteolysis of intercellular cohesive structures preceding desquamation, which is the shedding of the outermost layer of the epidermis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty