Would you like to log in again?
MMAE Antibody Drug Conjugate (ADC) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
KTR-757
102993-776EA 2909.62 USD InStock
102993-776
MMAE Antibody Drug Conjugate (ADC) ELISA, Eagle Biosciences
Assays ELISAs
Human MMAE Antibody Drug Conjugate EIA detects antibody-MMAE-conjugate level in test samples.
  • Dynamic Range: 0.032 - 4.0 µg/mL
  • Sensitivity: 0.0101 µg/ml
  • Incubation: 2.5 hours
The Eagle Biosciences Human MMAE Antibody Drug Conjugate (ADC) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for use in the quantitative determination of antibody-MMAE-conjugate level in test sample. It is useful for pre-clinical and clinical pharmacology study of MMAE Antibody Drug Conjugate (ADC).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty