Would you like to log in again?
Ubiquinone / Coenzyme Q10 HPLC Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
Q1031-H100
102993-784EA 1275 USD InStock
102993-784
Ubiquinone / Coenzyme Q10 HPLC Assay, Eagle Biosciences, Inc.
Assays Immunological Assays
The Ubiquinone (Coenzyme Q10) HPLC Assay kit is intended to determine ubiquinone in fluids.
  • Incubation Time: 10 minutes
  • Sample Size: 200 µl
  • Sample Type: Serum, Plasma, EDTA Whole Blood
Ubiquinone is a coenzyme of every cell in the body. Ubiquinone is made up by a chinonic ring and an isoprenic side chain. In humans ubiquinone can be synthesized and taken up by nutrition.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty