Would you like to log in again?
Alpha 1 Antitrypsin ELISA , Eagle Biosciences
Supplier: Eagle Biosciences
A1A35-K01
102993-856EA 935 USD InStock
102993-856
Alpha 1 Antitrypsin ELISA , Eagle Biosciences
Assays ELISAs
Alpha-1 Antitrypsin ELISA is intended to detect alpha-1 antitrypsin in serum, plasma and stool.
  • Dynamic Range: 3.3 - 90 ng/ml
  • Sensitivity: 1.5 ng/ml
  • Incubation: 2 hours
Alpha-1-Antitrypsin is a 52 kD glycoprotein, which is produced by the liver, intestinal macrophages, monocytes and mucous membrane cells of the gut. It belongs to the group of acute phase proteins and is one of the most important proteinase inhibitors. Alpha-1-antitrypsin inhibits, beside others, the proteinases trypsin and the elastase of neutrophils. A lack of α-1-AT leads to an enhanced proteolysis. Only a very small amount of alpha-1-antitrypsin is cleaved or resorbed in the gut. Therefore the measurement of α-1-AT in stool reflects the permeability of the gut during inflammatory processes. The Eagle Biosciences Alpha-1 Antitrypsin ELISA Kit allows an easy, rapid and precise quantitative determination of alpha-1-antitrypsin in biological samples. The kit includes all reagents ready to use for preparation of the samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty