Would you like to log in again?
ADMA (Asymmetric Dimethylarginine) / Arginine ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
EA207/192
102993-870EA 2602.31 USD InStock
102993-870
ADMA (Asymmetric Dimethylarginine) / Arginine ELISA, Eagle Biosciences, Inc.
Assays ELISAs
ADMA / Arginine ELISA detects Endogenous ADMA and L-Arginine in EDTA-Plasma, for research use only.
  • Dynamic Range: ADMA: 0.1 - 5.0 µmol/L, Arginine: 6 - 600 µmol/L
  • Sensitivity: ADMA: 0.01 µmol/L, Arginine: 3.0 µmol/L
  • Incubation: Overnight
Importantly, high ADMA levels and low L-arginine/ADMA ratio were both independent predictors of death in the community-based Framingham Offspring Study. As ADMA competes with L-arginine for binding to NO synthase, many scientists suggest that the L-arginine/ADMA ratio is a better index of NOS substrate availability and, thus, functional integrity of the NOS pathway, than L-arginine levels alone.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty