Would you like to log in again?
Anti-ASGPR (Asialoglycoprotein Receptor) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
3900
102993-872EA 568.85 USD InStock
102993-872
Anti-ASGPR (Asialoglycoprotein Receptor) ELISA, Eagle Biosciences
Assays ELISAs
Anti-ASGPR ELISA detects IgG antibodies to asialoglycoprotein receptor in human serum or plasma.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut Off Control
  • Incubation: 2 hours
Anti-ASGPR (Asialoglycoprotein Receptor) ELISA Assay Kit is used for the semi-quantitative determination of IgG antibodies to asialoglycoprotein receptor (ASGPR) in human serum or plasma. ASGPR is a liver specific membrane receptor playing a pivotal role in the endocytosis of glycoproteins from the blood. An induction of humoral and cellular immune mechanisms to the ASGPR has been observed in the course of inflammatory liver disorders especially autoimmune hepatitis. The level of ASGPR autoantibodies correlates with the severity of the disease and declines under therapy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty