Would you like to log in again?
Anti-beta2 adrenergic receptor antibodies (IgG) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
AB231-K01
102993-876EA 2720 USD InStock
102993-876
Anti-beta2 adrenergic receptor antibodies (IgG) ELISA, Eagle Biosciences
Assays ELISAs
Beta2 Adr Rcptr Ab IgG ELISA detects Ab against the beta2-adrenergic receptor in serum and plasma.
  • Dynamic Range: 2.5 - 40 U/ml
  • Sensitivity: 2.5 U/mL
  • Incubation: 3.5 hours
Introduction
The β1-adrenergic receptor is a G protein-coupled receptor. β receptors have the subtypes β1, β2 and β3. Agonist binding causes a rise in the intracellular concentration of the second messenger The β-2 adrenergic receptor is a G protein-coupled receptor. β receptors have the subtypes β1, β2 and β3. Agonist binding causes a rise in the intracellular concentration of the second messenger cAMP.
The Eagle Biosciences Human anti-β2-adrenergic receptor antibody EIA is designed for the determination of antibodies (IgG) against the β2-adrenergic receptor in serum and plasma.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty