Would you like to log in again?
Anti-AChR (Acetylcholine Receptor) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
3104
102993-878EA 2295 USD InStock
102993-878
Anti-AChR (Acetylcholine Receptor) ELISA, Eagle Biosciences
Assays ELISAs
Anti-AChR ELISA determinesAnti-Acetylcholine Receptor in serum.
  • Dynamic Range: 0.2 - 20 IU/ml
  • Sensitivity: 0.23 nmol/l
  • Incubation: overnight
The Eagle Biosciences Anti-AChR (Acetylcholine Receptor) ELISA Assay kit is intended for the quantitative determination of Anti-AChR in serum by enzyme linked immunoassay (ELISA). The Anti-AChR ELISA Assay kit is for research use only and not to be used in diagnostic procedures. Autoantibodies to the acetylcholine receptor (AChR) are responsible for failure of the neuromuscular junction in myasthenia gravis, a neuromuscular disease leading to fluctuating muscle weakness and fatigability. Measurement of these antibodies can be of considerable value in disease diagnosis and management.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty