Would you like to log in again?
Adrenocorticotropic Hormone (ACTH) ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
ACT31-K01
102993-880EA 621.15 USD InStock
102993-880
Adrenocorticotropic Hormone (ACTH) ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Human Adrenocorticotrophic Hormone (ACTH) ELISA is intended to detect human ACTH levels in plasma.
  • Dynamic Range: 7.6 - 416 pg/ml
  • Sensitivity: 0.4 pg/ml
  • Incubation: 2.5 hours
The Eagle Biosciences Human Adrenocorticotrophic Hormone (ACTH) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Adrenocorticotrophic Hormone (ACTH) levels in plasma. The Eagle Biosciences Human Adrenocorticotrophic Hormone (ACTH) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty