Would you like to log in again?
Albumin (Mouse) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
ALB11-K04 ALB11-K01 ALB11-K08
103389-042EA 4086.54 USD InStock
103389-042 102993-900 102993-902
Albumin (Mouse) ELISA, Eagle Biosciences
Assays ELISAs
Mouse Albumin ELISA is to detect albumin in mouse urine samples.
  • Dynamic Range: 2.5 to 10240 ng/ml
  • Sensitivity: 0.1 µg/ml
  • Incubation: 2 hours
This kit is designed for the quantitative determination of albumin in mouse urine samples.
Caution: For research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty