Would you like to log in again?
Albumin (Rat) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
ALB21-K08 ALB21-K04 ALB21-K01
102993-904EA 1340.38 USD InStock
102993-904 103389-040 102993-906
Albumin (Rat) ELISA, Eagle Biosciences
Assays ELISAs
Rat Albumin ELISA is to detect albumin in rat urine samples.
  • Dynamic Range: 10 to 10240 ng/ml
  • Sensitivity: 0.4 µg/ml
  • Incubation: 2 hours
This kit is designed for the quantitative determination of albumin in rat urine samples.
Caution: For research use only and should not be used in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty