Would you like to log in again?
Anti-M2 (Mitochondrial Antibodies) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4052
102993-912EA 778.08 USD InStock
102993-912
Anti-M2 (Mitochondrial Antibodies) ELISA, Eagle Biosciences
Assays ELISAs
Anti-M2 ELISA is to detect IgG antibodies to the mitochondrial antigen M2 in human serum or plasma.
  • Dynamic Range: 3 -300 U/ml
  • Sensitivity: 3 U/ml
  • Incubation: 2 hours
The group of primary autoimmune liver disease (PAL) comprises autoimmune hepatitis (AIH), PBC and primary sclerosing cholangitis (PSC). The clinical picture of PAL is in most cases not different from other chronic liver diseases. About 15% of all cases with chronic liver diseases show an autoimmune pathogenesis. Therefore, after exclusion of infectious etiology especially by viruses, the determination of different autoantibodies is recommended. PBC is a chronic inflammatory disorder of the small and medium bile ducts and serologically characterized by the occurrence of circulating M2 autoantibodies. M2 autoantibodies belong to the group of mitochondrial antibodies (AMA) and recognize three related proteins of the alpha-keto acid dehydrogenase complex. This complex is located at the inner mitochondrial membrane. The major epitopes of this protein complex is found on the E2 subunit and the protein X of the pyruvate dehydrogenase complex (PDC). Furthermore, M2 autoantibodies bind the E1alpha and E1beta subunits of the same complex and the E2 subunit of several other multi-enzyme complexes, such as the 2-oxo-glutarate dehydrogenase complex (OGDC) and the branched chain 2-oxo acid dehydrogenase complex (BCOADC).M2 autoantibodies have been found in up to 96% of patients suffering from PBC thus representing a powerful serological tool for the diagnosis of PBC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty