Would you like to log in again?
Adrenaline, Noradrenaline & Dopamine ELISA , Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
EA603/288
102993-918EA 1261.92 USD InStock
102993-918
Adrenaline, Noradrenaline & Dopamine ELISA , Eagle Biosciences, Inc.
Assays ELISAs
Adrenaline, Noradrenaline, Dopamine ELISA is detected in plasma and urine
  • Dynamic Range: Adrenaline: 0.5 - 250 ng/ml, Noradrenaline: 1.5 - 500 ng/ml, Dopamine:  1.5 - 2560 ng/ml
  • Sensitivity: URINE:  Adrenaline: 0.16 ng/ml, Noradrenaline: 0.85 ng/ml, Dopamine 0.46 ng/ml, PLASMA: Adrenaline: 0.8 pg/ml, Noradrenaline: 43 pg/ml, Dopamine 23 pg/ml
  • Incubation: Overnight
The Adrenaline, Noradrenaline, & Dopamine (Plasma & Urine) ELISA assay kit provides materials for the quantitative measurement of adrenaline, noradrenaline and dopamine in plasma and urine. Noradrenaline, adrenaline and dopamine are extracted using a cis-diol-specific affinity gel and acylated to N-acylnoradrenaline, N-acyladrenaline and N-acyl-dopamine and then converted enzymatically into N-acylnormeta­nephrine, N-acylmetanephrine and N-acyl-3-methoxytyramine.
For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty