Would you like to log in again?
ANA screen (Antinuclear Antibody) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4010
102993-922EA 568.85 USD InStock
102993-922
ANA screen (Antinuclear Antibody) ELISA, Eagle Biosciences
Assays ELISAs
ANAscreen detects autoantibodies to nuclear and cytoplasmic antigens in human serum and plasma.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut Off Control
  • Incubation: 2.5 hours
Systemic autoimmune diseases such as systemic lupus erythematosus, scleroderma, rheumatoid arthritis, Sjögren’s syndrome, dermatomyositis, mixed connective tissue disease are characterized by the appearance of a variety of autoantibodies directed against components of the cell nucleus. Although significance and pathological relevance of some auto-antibodies are not completely revealed yet, the detection of auto-antibodies is widely established and plays an important role in the diagnosis of systemic autoimmune diseases (1,2,3). ANAscreen allows the detection of total autoantibodies to nuclear and cytoplasmic Antigenss in one sample as a summary parameter in the diagnosis of systemic autoimmune disorders. Antigensic combination of complete HeLa nuclei with recombinant proteins and purified native Antigenss guarantees a maximum of sensitivy and specificity for the ANA detection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty