Would you like to log in again?
Rat Anti Angiotensin Receptor 1 IgM Antibody ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
ARM21-K01
102993-932EA 2720 USD InStock
102993-932
Rat Anti Angiotensin Receptor 1 IgM Antibody ELISA, Eagle Biosciences
Assays ELISAs
The Rat ATR1 IgM Antibody ELISA is for research use only and not for use in diagnostic procedures.
  • Dynamic Range: 1.25 -40 U/ml
  • Sensitivity: Cut-off
  • Incubation: 3.5 hours
Angiotensin II is a peptide hormone which causes vasoconstriction, increased blood pressure, and release of aldosterone from the adrenal cortex. It occupies an important role in the Renin-Angiotensin-System (RAS). Angiotensin II mediates the effects through G-Protein-coupled receptors, the Angiotensin receptors. The occurrence of autoantibodies against AT1 receptor is associated with an increased risk of an immunologic rejection after organ transplantation. The presence of AT1 autoantibodies correlate with the existence and course of Scleroderma.
This Rat Anti Angiotensin Receptor 1 IgM Antibody ELISA Assay is designed for the determination of Rat antibodies (IgM) against the Angiotensin II receptor subtype I in serum and plasma.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty