Would you like to log in again?
Anti Angiotensin Receptor 1 IgM Antibody ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
ARM31-K01
102993-934EA 2720 USD InStock
102993-934
Anti Angiotensin Receptor 1 IgM Antibody ELISA, Eagle Biosciences
Assays ELISAs
The Human ATR1 IgM Antibody ELISA is for research use only and not for diagnostic procedures.
  • Dynamic Range: 2.5 - 40 U/ml
  • Sensitivity: 2.5 U/mL
  • Incubation: 3.5 hours
Angiotensin II is a peptide hormone which causes vasoconstriction, increased blood pressure, and release of aldosterone from the adrenal cortex. It occupies an important role in the Renin-Angiotensin-System (RAS). Angiotensin II mediates the effects through G-Protein-coupled receptors, the Angiotensin receptors. The occurrence of autoantibodies against AT1 receptor is associated with an increased risk of an immunologic rejection after an organ transplantation. The presence of AT1 autoantibodies correlate with the existence and course of Scleroderma.
The Eagle Biosciences Human Anti Angiotensin Receptor 1 IgM Antibody Assay Kit anti-Angiotensin receptor 1-IgM-Antibody EIA is designed for the determination of IgM-Antibodies against the Angiotensin II receptor subtype I in serum and plasma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty