Would you like to log in again?
ASCA IgG ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4007
102993-940EA 568.85 USD InStock
102993-940
ASCA IgG ELISA, Eagle Biosciences
Assays ELISAs
ASCA IgG ELISA determines IgG antibodies to Saccharomyces cerevisiae in human serum.
  • Dynamic Range: 1-300 U/mL
  • Sensitivity: 1 U/mL
  • Incubation: 2.5 hours
Non-specific inflammatory bowel diseases includ-ing Crohn’s disease (Enteritis regionalis) and ul-cerative colitis (UC) are characterized by unknown etiology as well as chronic-remitting inflammatory processes of the intestine. Whereas the inflamma-tion of ulcerative colitis is restricted to the mucosa and submucosa of colon and rectum, Crohn’s dis-ease (CD) shows a wide spread inflammation of the gastro-intestinal tract with granuloma for-mation.  The risk developing one of these diseases is strongly influenced by immunologic, genetic, in-fectious and environmental factors. The differential diagnosis of inflammatory bowel diseases to chronic diarrhea, recurrent abdominal dolor, infectious colitis, anorexia as well as the differentiation of CD to ulcerative colitis is still a high challenge. The determination of IgA and IgG antibodies to Saccharomyces cerevisiae (baker’s yeast) has been described as one important serological marker for the differential diagnosis of Crohn’s disease re-cently. Up to 70 % of patients with CD show anti-body levels to Saccharomyces cerevisiae. Although the cause for their occurrence has been unclear, antibodies to Saccharomyces cerevisiae (ASCA) are strongly associated with inflammatory pro-cesses of the intestine. In combination with the detection of autoanti-bodies to atypical anti-neutrophil cytoplasmic Antigenss (aANCA) which are mainly found in patients with ulcerative colitis, ASCA are a valid pa-rameter for the differentiation of Crohn’s disease and ulcerative colitis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty