Would you like to log in again?
Anti-Angiotensin II Receptor 1 (AT1) Antibodies IgG (Rat) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
AT121-K01
102993-944EA 2818.08 USD InStock
102993-944
Anti-Angiotensin II Receptor 1 (AT1) Antibodies IgG (Rat) ELISA, Eagle Biosciences
Assays ELISAs
The Rat Anti Angiotensin Receptor 1 (ATR1) IgG Antibody ELISA is an antibody screening test.
  • Sensitivity: Cut-off
  • Dynamic Range: 1.25 - 40 U/mL
  • Incubation Time: 3.5 hours
Angiotensin II is a peptide hormone which causes vasoconstriction, increased blood pressure, and release of aldosterone from the adrenal cortex. It occupies an important role in the ReninAngiotensin-System (RAS). Angiotensin II mediates the effects through G-Protein-coupled receptors, the Angiotensin receptors. The occurrence of autoantibodies against AT1 receptor is associated with an increased risk of an immunologic rejection after organ transplantation. The presence of AT1 autoantibodies correlate with the existence and course of Scleroderma. This Rat Anti Angiotensin II Receptor 1 (AT1R) IgG Antibody ELISA Assay Kit is designed for the determination of Rat antibodies (IgG) against the Angiotensin II receptor subtype I in serum and plasma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty