Would you like to log in again?
Anti-ß2 GP-1 screen (Beta 2 Glycoprotein 1) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4036
102993-950EA 725.77 USD InStock
102993-950
Anti-ß2 GP-1 screen (Beta 2 Glycoprotein 1) ELISA, Eagle Biosciences
Assays ELISAs
Anti-Beta 2 GP-I Screen ELISA determines IgG,IgM and IgA Ab (screening) to Beta2 glycoprotein-I.
  • Dynamic Range: 10-300 U/mL
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours
APAS is an autoimmune disorder comprising such clinical symptoms like arterial or venous thrombosis, thrombocytopenia and recurrent fetal loss. Primary APAS as well as systemic lupus erythematosus (SLE) are characterized by the appearance of autoantibodies to negatively charged phospholipids (1). Although significance and pathological relevance of phospholipid anti¬bodies are not completely revealed yet, the detection of several autoantibody specificities is usually applied to the differential diagnosis and follow-up of systemic rheumatic inflammatory diseases. Unlike phospholipid antibodies which occur in some patients having infectious disease, phospholipid antibodies of autoimmune disease patients seem to recognize the relevant phospholipids in association with a plasma protein cofactor. One of these cofactors has been identified as 2 glycoprotein-I (2 GP-I) (apo¬lipo¬pro¬tein H) (2,3). 2 GP-I, a serum protein with a molecular weight of 50 kDa affects platelet aggregation and coagulation. The positively charged fifth domain of 2 GP-I interacts with negatively charged phospholipids or activated polystyrol surfaces of ELISA wells. This interaction results in conformational changes of 2 GP-I and the creation of new epitopes apparently recognized by autoimmune phospholipid autoantibodies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty